Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302652.1 | NP_001289581.1 | 5 | 593 | ||
NM_001302653.1 | NP_001289582.1 | 6 | 683 | |||
NM_032374.4 | NP_115750.2 | 5 | 595 | |||
NR_126431.1 | 6 | 760 | ||||
NR_126432.1 | 4 | 531 | ||||
GenBank mRNA | AK223092.1 | - | 5 | 566 | ||
AK223103.1 | - | 5 | 566 | |||
BC007412.2 | - | 5 | 558 | 2989951 | ||
BF528062.1 | - | 6 | 661 | |||
BP204024.1 | - | 3 | 319 | |||
BU956674.1 | - | 4 | 488 | |||
BX340258.2 | - | 4 | 510 | |||
HY134136.1 | - | 3 | 334 |