Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHEK2 SIRNA Assays ›
Gene Symbol:
CHEK2
Gene Name:
checkpoint kinase 2
Gene Aliases:
CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1
Chromosome Location:
Chr.22: 28687743 - 28741866 on Build GRCh38
UniGene ID:
Hs.291363
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533980
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHEK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005735.1
NP_001005735.1
8
1009
NM_001257387.1
NP_001244316.1
8
994
NM_007194.3
NP_009125.1
7
880
NM_145862.2
NP_665861.1
7
880
XM_006724114.3
XP_006724177.2
3
523
XM_006724116.2
XP_006724179.2
3
268
XM_011529839.2
XP_011528141.1
8
1031
XM_011529840.2
XP_011528142.1
8
1033
XM_011529841.1
XP_011528143.1
7
812
XM_011529842.2
XP_011528144.1
6
700
XM_011529843.1
XP_011528145.1
6
679
XM_011529844.2
XP_011528146.1
8
1041
XM_011529845.2
XP_011528147.1
6
587
XM_017028560.1
XP_016884049.1
8
995
XM_017028561.1
XP_016884050.1
4
239
GenBank mRNA
AB040105.1
-
6
808
AF086904.1
-
6
808
AF096279.1
-
6
808
AF174135.1
-
6
808
AF217975.1
-
8
1593
AJ131197.1
-
6
808
AK289360.1
-
7
859
AK290754.1
-
7
882
AY551297.1
-
7
937
AY551298.1
-
6
808
AY551299.1
-
6
808
AY551300.1
-
6
808
AY551301.1
-
6
698
AY551302.1
-
5
717
AY551303.1
-
4
535
AY551305.1
-
7
922
BC004207.2
-
7
835
3532866
CR456418.1
-
8
1008
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
G2/M transition of mitotic cell cycle
replicative cell aging
positive regulation of protein phosphorylation
double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
intrinsic apoptotic signaling pathway in response to DNA damage
response to gamma radiation
peptidyl-serine phosphorylation
cellular response to drug
regulation of protein catabolic process
signal transduction in response to DNA damage
cellular protein catabolic process
positive regulation of transcription, DNA-templated
protein autophosphorylation
protein stabilization
cell division
negative regulation of cell cycle arrest
signal transduction involved in intra-S DNA damage checkpoint
mitotic spindle assembly
replicative senescence
regulation of signal transduction by p53 class mediator
response to doxorubicin
cellular response to bisphenol A
negative regulation of DNA damage checkpoint
positive regulation of anoikis
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0