Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIM37 SIRNA Assays ›
          Gene Symbol:
          TRIM37
          Gene Name:
          tripartite motif containing 37
          Gene Aliases:
          MUL, POB1, TEF3
          Chromosome Location:
          Chr.17: 58968068 - 59106921 on Build GRCh38
          UniGene ID:
          Hs.579079
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534002

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIM37
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005207.3 NP_001005207.1 16 2096
          NM_001320987.1 NP_001307916.1 15 1994
          NM_001320988.1 NP_001307917.1 16 2096
          NM_001320989.1 NP_001307918.1 16 2096
          NM_001320990.1 NP_001307919.1 17 2172
          NM_015294.4 NP_056109.1 16 2096
          XM_005257385.1 XP_005257442.1 16 1928
          XM_011524831.1 XP_011523133.1 16 1928
          XM_011524832.2 XP_011523134.1 16 1928
          XM_011524833.1 XP_011523135.1 15 1877
          XM_011524834.1 XP_011523136.1 15 1810
          XM_011524836.1 XP_011523138.1 16 1928
          XM_017024662.1 XP_016880151.1 16 1928
          XM_017024663.1 XP_016880152.1 16 1928
          XM_017024664.1 XP_016880153.1 15 1877
          XM_017024665.1 XP_016880154.1 15 1810
          XM_017024666.1 XP_016880155.1 15 1877
          XM_017024667.1 XP_016880156.1 15 1877
          XM_017024668.1 XP_016880157.1 15 1810
          XM_017024669.1 XP_016880158.1 14 1759
          XM_017024670.1 XP_016880159.1 15 1809
          XM_017024671.1 XP_016880160.1 15 1840
          XM_017024672.1 XP_016880161.1 15 1840
          XM_017024673.1 XP_016880162.1 15 1632
          XM_017024674.1 XP_016880163.1 15 1632
          GenBank mRNA AB020705.1 - 16 1681
          AF213365.1 - 16 1900
          AK025648.1 - 10 1114
          AK289674.1 - 16 1911
          AK292459.1 - 16 1968
          AK294850.1 - 15 1899
          BC036012.1 - 16 1932 5270430
          BX537955.1 - 17 1991

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of NF-kappaB transcription factor activity
          histone H2A monoubiquitination
          histone H2A-K119 monoubiquitination
          negative regulation of centriole replication
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein autoubiquitination
          aggresome assembly
          chromatin binding
          ubiquitin-protein transferase activity
          tumor necrosis factor receptor binding
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase binding
          protein homodimerization activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0