Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534009
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
21
2728
NM_001145848.1
NP_001139320.1
21
2444
NM_001145849.1
NP_001139321.1
21
2448
NM_001145850.1
NP_001139322.1
21
2448
NM_001145851.1
NP_001139323.1
20
2421
NM_001145852.1
NP_001139324.1
20
2421
NM_006017.2
NP_006008.1
21
2448
XM_005248195.4
XP_005248252.1
21
2544
XM_005248196.4
XP_005248253.1
21
2544
XM_006713974.3
XP_006714037.1
20
2049
XM_011513890.1
XP_011512192.1
22
2530
XM_011513892.2
XP_011512194.1
22
2681
XM_011513893.2
XP_011512195.1
22
2755
XM_011513894.2
XP_011512196.1
22
2840
XM_011513895.2
XP_011512197.1
23
2956
XM_011513896.2
XP_011512198.1
22
2585
XM_011513897.2
XP_011512199.1
22
2772
XM_011513898.2
XP_011512200.2
22
2530
XM_011513899.2
XP_011512201.2
21
2544
XM_011513900.2
XP_011512202.1
22
2530
XM_011513902.2
XP_011512204.1
22
2530
XM_011513903.2
XP_011512205.1
21
2079
XM_017008799.1
XP_016864288.1
21
2574
XM_017008800.1
XP_016864289.1
22
2530
XM_017008801.1
XP_016864290.1
22
2530
XM_017008802.1
XP_016864291.1
22
2622
XM_017008803.1
XP_016864292.1
21
2519
XM_017008804.1
XP_016864293.1
21
2589
XM_017008805.1
XP_016864294.1
21
2462
GenBank mRNA
AF027208.1
-
21
2273
AF117225.1
-
21
2489
AF507034.1
-
20
2246
AK027420.1
-
21
2467
AK027422.1
-
21
2489
AK075411.1
-
21
2444
AK226033.1
-
21
2467
AK314529.1
-
21
2488
AY449689.1
-
20
2209
AY449690.1
-
20
2209
AY449691.1
-
20
2209
AY449692.1
-
21
2236
AY449693.1
-
21
2236
BC012089.1
-
21
2444
4644690
HQ628626.1
-
20
2209
HQ628627.1
-
22
2465
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0