Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GOSR2 SIRNA Assays ›
Gene Symbol:
GOSR2
Gene Name:
golgi SNAP receptor complex member 2
Gene Aliases:
Bos1, EPM6, GS27
Chromosome Location:
Chr.17: 46923086 - 46975519 on Build GRCh38
UniGene ID:
Hs.463278
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534163
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GOSR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012511.2
NP_001012529.1
3
189
NM_001321133.1
NP_001308062.1
3
189
NM_001321134.1
NP_001308063.1
3
237
NM_004287.4
NP_004278.2
3
189
NM_054022.3
NP_473363.1
3
189
XM_005257844.3
XP_005257901.1
3
198
XM_006722190.3
XP_006722253.1
3
198
XM_011525501.2
XP_011523803.1
3
198
XM_011525502.2
XP_011523804.1
3
198
XM_017025378.1
XP_016880867.1
3
195
XM_017025379.1
XP_016880868.1
3
189
XM_017025380.1
XP_016880869.1
3
546
XM_017025381.1
XP_016880870.1
3
172
XM_017025382.1
XP_016880871.1
3
524
XM_017025383.1
XP_016880872.1
3
198
XM_017025384.1
XP_016880873.1
3
220
XM_017025385.1
XP_016880874.1
3
195
XM_017025386.1
XP_016880875.1
3
198
XM_017025387.1
XP_016880876.1
3
195
XM_017025388.1
XP_016880877.1
3
198
XM_017025389.1
XP_016880878.1
3
205
XM_017025390.1
XP_016880879.1
3
550
XM_017025391.1
XP_016880880.1
3
548
XM_017025392.1
XP_016880881.1
3
198
XM_017025393.1
XP_016880882.1
3
195
XM_017025394.1
XP_016880883.1
3
546
XM_017025395.1
XP_016880884.1
3
546
GenBank mRNA
AF007548.1
-
3
113
AF229796.1
-
3
144
AK290890.1
-
3
164
BC009710.2
-
3
160
3890308
BC034762.1
-
3
132
5185659
BG718145.1
-
3
183
BM846811.1
-
3
183
CD048223.1
-
3
214
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
IRE1-mediated unfolded protein response
retrograde transport, endosome to Golgi
COPII vesicle coating
vesicle fusion with Golgi apparatus
membrane fusion
SNARE binding
transporter activity
SNAP receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0