Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VPS13B SIRNA Assays ›
          Gene Symbol:
          VPS13B
          Gene Name:
          vacuolar protein sorting 13 homolog B
          Gene Aliases:
          CHS1, COH1
          Chromosome Location:
          Chr.8: 99007014 - 99877586 on Build GRCh38
          UniGene ID:
          Hs.191540
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534184

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VPS13B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015243.2 NP_056058.2 7 1014
          NM_017890.4 NP_060360.3 7 1014
          NM_152564.4 NP_689777.3 7 1014
          NM_181661.2 NP_858047.2 7 1014
          NR_047582.1 8 1129
          XM_005250800.3 XP_005250857.1 7 1320
          XM_005250801.4 XP_005250858.1 7 7190
          XM_006716510.3 XP_006716573.1 7 1006
          XM_011516848.2 XP_011515150.1 7 1006
          XM_011516849.2 XP_011515151.1 7 1006
          XM_011516850.2 XP_011515152.1 5 646
          XM_011516853.2 XP_011515155.1 7 1006
          XM_011516859.2 XP_011515161.1 7 1006
          XM_011516861.1 XP_011515163.1 7 1006
          XM_011516865.1 XP_011515167.1 7 1006
          XM_011516866.2 XP_011515168.1 7 1006
          XM_017013109.1 XP_016868598.1 7 1006
          GenBank mRNA AJ608772.1 - 7 1006
          AJ608773.1 - 7 1006
          AK000590.1 - 7 1040
          AK223122.1 - 7 1040
          AY223814.1 - 7 1014
          AY223815.1 - 7 1014
          AY223816.1 - 7 1014
          AY223817.1 - 7 1014
          AY223818.1 - 7 1014
          BC094749.1 - 7 970
          BC132976.1 - 6 937
          BC132978.1 - 6 937

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s)

          protein transport

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0