Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSPP1 SIRNA Assays ›
          Gene Symbol:
          CSPP1
          Gene Name:
          centrosome and spindle pole associated protein 1
          Gene Aliases:
          CSPP, JBTS21
          Chromosome Location:
          Chr.8: 67062417 - 67196614 on Build GRCh38
          UniGene ID:
          Hs.370147
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534257

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSPP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291339.1 NP_001278268.1 26 3550
          NM_024790.6 NP_079066.5 29 3550
          XM_005251305.3 XP_005251362.2 32 4034
          XM_006716474.2 XP_006716537.2 31 3881
          XM_006716477.2 XP_006716540.2 27 3503
          XM_011517598.1 XP_011515900.1 33 4076
          XM_011517599.1 XP_011515901.1 32 4052
          XM_011517600.1 XP_011515902.1 31 4010
          XM_011517601.1 XP_011515903.1 31 3971
          XM_011517602.1 XP_011515904.1 30 3929
          XM_011517603.1 XP_011515905.1 32 3759
          XM_011517607.1 XP_011515909.1 31 3639
          XM_011517608.1 XP_011515910.1 30 3636
          XM_011517609.1 XP_011515911.1 29 3758
          XM_011517611.2 XP_011515913.1 17 2135
          XM_017013847.1 XP_016869336.1 31 3935
          XM_017013848.1 XP_016869337.1 30 3911
          XM_017013849.1 XP_016869338.1 30 3872
          XM_017013850.1 XP_016869339.1 29 3830
          XM_017013851.1 XP_016869340.1 29 3594
          XM_017013852.1 XP_016869341.1 28 3677
          XM_017013853.1 XP_016869342.1 28 3441
          XM_017013854.1 XP_016869343.1 26 3479
          XM_017013855.1 XP_016869344.1 24 3245
          XM_017013856.1 XP_016869345.1 24 3063
          XM_017013857.1 XP_016869346.1 25 3113
          XM_017013858.1 XP_016869347.1 23 2861
          XM_017013859.1 XP_016869348.1 19 2263
          GenBank mRNA AJ583433.1 - 26 3550
          AK026143.1 - 997
          AK308372.1 - 12 1386
          AM156947.1 - 29 3551
          BC022867.1 - 11 1049
          BX649147.1 - 29 3552

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          centromere DNA-binding protein

          Gene Ontology Categories:

          Function(s)

          positive regulation of cytokinesis
          positive regulation of cell division

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0