Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534258
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
24, 25
3338
NM_024790.6
NP_079066.5
27, 28
3338
XM_005251305.3
XP_005251362.2
30, 31
3822
XM_006716474.2
XP_006716537.2
29, 30
3669
XM_006716477.2
XP_006716540.2
25, 26
3291
XM_011517598.1
XP_011515900.1
31, 32
3864
XM_011517599.1
XP_011515901.1
30, 31
3840
XM_011517600.1
XP_011515902.1
29, 30
3798
XM_011517601.1
XP_011515903.1
29, 30
3759
XM_011517602.1
XP_011515904.1
28, 29
3717
XM_011517603.1
XP_011515905.1
30, 31
3547
XM_011517607.1
XP_011515909.1
29, 30
3427
XM_011517608.1
XP_011515910.1
28, 29
3424
XM_011517609.1
XP_011515911.1
27, 28
3546
XM_011517611.2
XP_011515913.1
15, 16
1923
XM_017013847.1
XP_016869336.1
29, 30
3723
XM_017013848.1
XP_016869337.1
28, 29
3699
XM_017013849.1
XP_016869338.1
28, 29
3660
XM_017013850.1
XP_016869339.1
27, 28
3618
XM_017013851.1
XP_016869340.1
27, 28
3382
XM_017013852.1
XP_016869341.1
26, 27
3465
XM_017013853.1
XP_016869342.1
26, 27
3229
XM_017013854.1
XP_016869343.1
24, 25
3267
XM_017013855.1
XP_016869344.1
22, 23
3033
XM_017013856.1
XP_016869345.1
22, 23
2851
XM_017013857.1
XP_016869346.1
23, 24
2901
XM_017013858.1
XP_016869347.1
21, 22
2649
XM_017013859.1
XP_016869348.1
17, 18
2051
GenBank mRNA
AJ583433.1
-
24, 25
3338
AK026143.1
-
785
AM156947.1
-
27, 28
3339
BX649147.1
-
27, 28
3340
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bz4hs:80/100.66.77.47:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0