Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
ANKRD36C
Gene Name:
ankyrin repeat domain 36
ankyrin repeat domain 36C
Gene Aliases:
UNQ2430
-
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 95849548 - 95992320 on Build GRCh38
UniGene ID:
Hs.541894
Hs.646318
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s53436
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
30
2246
XM_006712514.3
XP_006712577.1
30
2556
XM_011511130.1
XP_011509432.1
30
2556
XM_011511131.1
XP_011509433.1
30
2556
XM_017004009.1
XP_016859498.1
30
2556
XM_017004010.1
XP_016859499.1
30
2556
XM_017004011.1
XP_016859500.1
30
2556
XM_017004012.1
XP_016859501.1
30
2556
XM_017004013.1
XP_016859502.1
30
2556
XM_017004014.1
XP_016859503.1
30
2556
XM_017004015.1
XP_016859504.1
28
2454
XM_017004016.1
XP_016859505.1
30
2556
XM_017004017.1
XP_016859506.1
30
2556
XM_017004018.1
XP_016859507.1
28
2454
XM_017004020.1
XP_016859509.1
30
2556
XM_017004021.1
XP_016859510.1
30
2556
XM_017004022.1
XP_016859511.1
28
2454
XM_017004023.1
XP_016859512.1
30
2556
XM_017004024.1
XP_016859513.1
29
2418
XM_017004025.1
XP_016859514.1
30
2556
XM_017004026.1
XP_016859515.1
30
2556
XM_017004027.1
XP_016859516.1
30
2556
XM_017004028.1
XP_016859517.1
30
2556
XM_017004029.1
XP_016859518.1
30
2556
XM_017004030.1
XP_016859519.1
30
2556
XM_017004031.1
XP_016859520.1
30
2556
XM_017004032.1
XP_016859521.1
30
2556
XM_017004033.1
XP_016859522.1
30
2556
XM_017004034.1
XP_016859523.1
30
2556
XM_017004036.1
XP_016859525.1
30
2556
XM_017004037.1
XP_016859526.1
30
2556
XM_017004038.1
XP_016859527.1
30
2556
XM_017004039.1
XP_016859528.1
30
2556
XM_017004040.1
XP_016859529.1
30
2556
XM_017004041.1
XP_016859530.1
30
2556
XM_017004042.1
XP_016859531.1
30
2556
XM_017004043.1
XP_016859532.1
30
2556
XM_017004044.1
XP_016859533.1
30
2556
XM_017004045.1
XP_016859534.1
30
2556
XM_017004046.1
XP_016859535.1
28
2311
XM_017004047.1
XP_016859536.1
27
2243
XM_017004048.1
XP_016859537.1
26
2170
XM_017004049.1
XP_016859538.1
28
2345
XM_017004050.1
XP_016859539.1
13
749
XM_017004051.1
XP_016859540.1
11
963
GenBank mRNA
AK307629.1
-
26
1767
BX647769.1
-
29
2118
DW413497.1
-
6
285
Gene Symbol: ANKRD36C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001310154.1
NP_001297083.1
32
2716
GenBank mRNA
AL832836.1
-
13
616
AL832977.1
-
5
181
Back To Top
More Information
Gene Ontology Categories:
Process(es)
ion channel inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0