Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534438
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
2
201
NM_001195249.1
NP_001182178.1
2
370
NM_001195250.1
NP_001182179.1
2
201
NM_001195251.1
NP_001182180.1
3
307
NM_001195252.1
NP_001182181.1
2
201
NM_001195254.1
NP_001182183.1
2
223
NM_175069.2
NP_778239.1
2
201
NM_175073.2
NP_778243.1
3
307
NR_036576.1
2
201
NR_036577.1
2
201
NR_036578.1
2
223
NR_036579.1
2
370
XM_006716791.3
XP_006716854.1
2
209
XM_006716792.2
XP_006716855.1
2
232
XM_011517938.1
XP_011516240.1
2
232
XM_011517939.2
XP_011516241.1
2
187
XM_017014831.1
XP_016870320.1
2
201
XM_017014832.1
XP_016870321.1
2
201
XM_017014833.1
XP_016870322.1
2
201
XM_017014834.1
XP_016870323.1
2
201
XM_017014835.1
XP_016870324.1
2
201
XM_017014836.1
XP_016870325.1
2
201
XM_017014837.1
XP_016870326.1
2
452
XM_017014838.1
XP_016870327.1
2
188
GenBank mRNA
AB603741.1
-
1
97
AJ565850.1
-
2
179
AJ565851.1
-
2
179
AJ565852.1
-
2
179
AJ565853.1
-
1
182
AJ565854.1
-
1
182
AJ565855.1
-
1
182
AJ575566.1
-
1
182
AK000164.1
-
1
319
AK055672.1
-
2
188
AK131046.1
-
2
153
AY040777.1
-
2
176
AY208829.1
-
2
176
AY208830.1
-
2
201
AY208831.1
-
2
167
AY208832.1
-
2
176
AY208833.1
-
2
176
AY208834.1
-
2
172
AY208835.1
-
2
161
AY208836.1
-
2
316
AY208837.1
-
3
273
AY208838.1
-
1
247
AY208839.1
-
2
315
AY208840.1
-
2
178
AY208841.1
-
1
130
AY302067.1
-
1
319
AY302068.1
-
1
319
AY302069.1
-
1
319
AY302070.1
-
1
319
AY302071.1
-
1
319
AY302072.1
-
1
319
AY302073.1
-
1
319
AY302074.1
-
1
319
BC001628.1
-
1
139
BC032650.1
-
2
247
5505120
BC068107.1
-
3
282
BC104881.1
-
2
215
8143884
BQ957536.1
-
3
290
BX538161.1
-
3
286
DB042208.1
-
2
223
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0