Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534439
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
4, 5
556
NM_001195249.1
NP_001182178.1
4, 5
725
NM_001195250.1
NP_001182179.1
3, 4
394
NM_001195251.1
NP_001182180.1
5, 6
662
NM_001195254.1
NP_001182183.1
3, 4
416
NM_175069.2
NP_778239.1
4, 5
556
NM_175073.2
NP_778243.1
5, 6
662
NR_036576.1
4, 5
551
NR_036577.1
4, 5
441
NR_036578.1
4, 5
573
NR_036579.1
4, 5
720
XM_006716791.3
XP_006716854.1
4, 5
564
XM_006716792.2
XP_006716855.1
4, 5
582
XM_011517938.1
XP_011516240.1
3, 4
540
XM_011517939.2
XP_011516241.1
3, 4
495
XM_017014831.1
XP_016870320.1
4, 5
556
XM_017014832.1
XP_016870321.1
4, 5
556
XM_017014833.1
XP_016870322.1
4, 5
556
XM_017014834.1
XP_016870323.1
4, 5
556
XM_017014835.1
XP_016870324.1
4, 5
556
XM_017014836.1
XP_016870325.1
4, 5
556
XM_017014837.1
XP_016870326.1
3, 4
645
XM_017014838.1
XP_016870327.1
4, 5
538
GenBank mRNA
AB603741.1
-
3, 4
452
AJ565850.1
-
4, 5
414
AJ565851.1
-
4, 5
419
AJ565852.1
-
4, 5
529
AJ565853.1
-
3, 4
532
AJ565854.1
-
3, 4
537
AJ565855.1
-
2, 3
375
AJ575566.1
-
3, 4
417
AK000164.1
-
3, 4
559
AK055672.1
-
4, 5
543
AK131046.1
-
4, 5
508
AY040777.1
-
4, 5
531
AY208829.1
-
4, 5
531
AY208830.1
-
3, 4
394
AY208832.1
-
3, 4
369
AY208833.1
-
3, 4
369
AY208835.1
-
4, 5
511
AY208836.1
-
4, 5
671
AY208837.1
-
5, 6
628
AY208838.1
-
3, 4
487
AY208839.1
-
4, 5
665
AY208840.1
-
4, 5
528
AY208841.1
-
3, 4
365
AY208842.1
-
1, 2
165
AY302067.1
-
3, 4
674
AY302068.1
-
3, 4
669
AY302069.1
-
3, 4
559
AY302070.1
-
3, 4
554
AY302071.1
-
3, 4
674
AY302072.1
-
3, 4
669
AY302073.1
-
3, 4
559
AY302074.1
-
3, 4
554
BC001628.1
-
3, 4
379
BC032650.1
-
4, 5
597
5505120
BC068107.1
-
5, 6
637
BC104881.1
-
4, 5
570
8143884
BQ957536.1
-
5, 6
645
BX538161.1
-
5, 6
641
DB042208.1
-
3, 4
416
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0