Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JAG1 SIRNA Assays ›
          Gene Symbol:
          JAG1
          Gene Name:
          jagged 1
          Gene Aliases:
          AGS, AHD, AWS, CD339, HJ1, JAGL1
          Chromosome Location:
          Chr.20: 10637684 - 10674046 on Build GRCh38
          UniGene ID:
          Hs.224012
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534506

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JAG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000214.2 NP_000205.1 14 2363
          XM_017027707.1 XP_016883196.1 13 2468
          GenBank mRNA AF003837.1 - 14 2306
          AF028593.1 - 14 2110
          AK302554.1 - 12 1713
          AK312722.1 - 14 1932
          BC098393.1 - 14 2266 30528888
          BC126205.1 - 15 1871 8991921
          BC126207.1 - 14 2099 8991923
          U61276.1 - 14 2217
          U73936.1 - 14 2260
          U77720.1 - 14 1809

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: L38483.
          This siRNA also targets the following transcripts in Mouse: AK137092, AF171092, BC058675.

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell fate determination
          blood vessel remodeling
          morphogenesis of an epithelial sheet
          T cell mediated immunity
          pulmonary valve morphogenesis
          cardiac right ventricle morphogenesis
          Notch signaling pathway
          Notch receptor processing
          nervous system development
          hemopoiesis
          keratinocyte differentiation
          regulation of cell migration
          response to muramyl dipeptide
          aorta morphogenesis
          regulation of cell proliferation
          auditory receptor cell differentiation
          myoblast differentiation
          endothelial cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of myeloid cell differentiation
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of Notch signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          cardiac septum morphogenesis
          ciliary body morphogenesis
          pulmonary artery morphogenesis
          cardiac neural crest cell development involved in outflow tract morphogenesis
          Notch signaling involved in heart development
          endocardial cushion cell development
          nephron development
          glomerular visceral epithelial cell development
          distal tubule development
          loop of Henle development
          neuronal stem cell population maintenance
          negative regulation of stem cell differentiation
          Notch binding
          structural molecule activity
          calcium ion binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0