siRNA Details
Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021926.3 | NP_068745.2 | 4 | 1691 | ||
GenBank mRNA | AB058691.1 | - | 4 | 1594 | ||
AK092346.1 | - | 5 | 1624 |

Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021926.3 | NP_068745.2 | 4 | 1691 | ||
GenBank mRNA | AB058691.1 | - | 4 | 1594 | ||
AK092346.1 | - | 5 | 1624 |