Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 2 | 297 | ||
XM_017007176.1 | XP_016862665.1 | 3 | 419 | |||
GenBank mRNA | AK027757.1 | - | 2 | 223 | ||
AK027877.1 | - | 2 | 161 | |||
AK075010.1 | - | 2 | 148 | |||
AK225589.1 | - | 2 | 220 | |||
AL136916.1 | - | 2 | 191 | |||
AY358625.1 | - | 2 | 60 | |||
BC003125.1 | - | 2 | 192 | 3503636 | ||
BC011719.2 | - | 2 | 192 | 4079766 | ||
DA259919.1 | - | 2 | 302 | |||
KU178769.1 | - | 1 | 43 | |||
KU178770.1 | - | 1 | 43 |