Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDN1 SIRNA Assays ›
          Gene Symbol:
          EDN1
          Gene Name:
          endothelin 1
          Gene Aliases:
          ARCND3, ET1, HDLCQ7, PPET1, QME
          Chromosome Location:
          Chr.6: 12256464 - 12297194 on Build GRCh38
          UniGene ID:
          Hs.511899
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534832

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EDN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168319.1 NP_001161791.1 3 623
          NM_001955.4 NP_001946.3 3 626
          XM_011514330.2 XP_011512632.1 4 382
          XM_011514331.2 XP_011512633.1 5 543
          XM_011514332.2 XP_011512634.1 4 380
          XM_017010331.1 XP_016865820.1 4 566
          GenBank mRNA AK226096.1 - 3 541
          AK291838.1 - 3 563
          AK313914.1 - 3 558
          BC009720.1 - 3 495 3900704
          BX439515.2 - 3 586
          CR541903.1 - 3 292
          S56805.1 - 3 628
          Y00749.1 - 3 544

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          prostaglandin biosynthetic process
          patterning of blood vessels
          in utero embryonic development
          histamine secretion
          regulation of systemic arterial blood pressure by endothelin
          regulation of pH
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          cell-cell signaling
          heart development
          respiratory gaseous exchange
          body fluid secretion
          positive regulation of cell proliferation
          dorsal/ventral pattern formation
          response to ozone
          multicellular organism aging
          positive regulation of heart rate
          positive regulation of endothelial cell migration
          positive regulation of cardiac muscle hypertrophy
          positive regulation of receptor biosynthetic process
          neural crest cell development
          phosphatidylinositol 3-kinase signaling
          response to activity
          artery smooth muscle contraction
          vein smooth muscle contraction
          glucose transport
          cell growth
          regulation of vasoconstriction
          sensory perception of pain
          calcium-mediated signaling
          peptide hormone secretion
          nitric oxide transport
          negative regulation of blood coagulation
          positive regulation of cell migration
          neutrophil chemotaxis
          negative regulation of cAMP biosynthetic process
          phospholipase D-activating G-protein coupled receptor signaling pathway
          negative regulation of cellular protein metabolic process
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          response to testosterone
          negative regulation of smooth muscle cell apoptotic process
          response to prostaglandin F
          response to nicotine
          cellular response to drug
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          response to muscle stretch
          epithelial fluid transport
          vasoconstriction
          protein kinase C deactivation
          middle ear morphogenesis
          positive regulation of odontogenesis
          superoxide anion generation
          rhythmic excitation
          response to amino acid
          positive regulation of MAP kinase activity
          positive regulation of JUN kinase activity
          response to leptin
          leukocyte activation
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell size
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle contraction
          positive regulation of hormone secretion
          negative regulation of hormone secretion
          inositol phosphate-mediated signaling
          positive regulation of smooth muscle cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          cartilage development
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          negative regulation of nitric-oxide synthase biosynthetic process
          membrane depolarization
          regulation of sensory perception of pain
          maternal process involved in parturition
          positive regulation of sarcomere organization
          positive regulation of prostaglandin-endoperoxide synthase activity
          positive regulation of cell growth involved in cardiac muscle cell development
          positive regulation of chemokine-mediated signaling pathway
          cellular response to calcium ion
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          cellular response to glucocorticoid stimulus
          cellular response to mineralocorticoid stimulus
          cellular response to fatty acid
          cellular response to hypoxia
          response to dexamethasone
          cellular response to transforming growth factor beta stimulus
          response to salt
          cytokine activity
          hormone activity
          protein binding
          endothelin A receptor binding
          endothelin B receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0