Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001701.3 | NP_001001701.2 | 1 | 366 | ||
NM_001170330.1 | NP_001163801.1 | 2 | 487 | |||
GenBank mRNA | AF075061.1 | - | 1 | 174 | ||
AK025703.1 | - | 1 | 130 | |||
AY605045.1 | - | 2 | 487 | |||
BC017399.1 | - | 1 | 189 | 4108498 | ||
DA353224.1 | - | 1 | 366 |