Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101676.1 | NP_001095146.1 | 2 | 211 | ||
NM_207506.2 | NP_997389.2 | 2 | 211 | |||
NR_109794.1 | 2 | 126 | ||||
XM_011517031.2 | XP_011515333.1 | 2 | 251 | |||
XM_011517034.2 | XP_011515336.1 | 2 | 250 | |||
XM_011517035.2 | XP_011515337.1 | 2 | 250 | |||
XM_017013409.1 | XP_016868898.1 | 2 | 253 | |||
XM_017013410.1 | XP_016868899.1 | 2 | 251 | |||
XM_017013411.1 | XP_016868900.1 | 2 | 251 | |||
XM_017013412.1 | XP_016868901.1 | 2 | 251 | |||
GenBank mRNA | AK096777.1 | - | 2 | 147 | ||
AK311704.1 | - | 2 | 126 | |||
BC121818.1 | - | 2 | 144 | 40109130 | ||
BC124554.1 | - | 2 | 144 | 40109129 | ||
DA459013.1 | - | 2 | 190 | |||
DB228621.1 | - | 2 | 126 |