Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BRCA1 SIRNA Assays ›
          Gene Symbol:
          BRCA1
          Gene Name:
          BRCA1, DNA repair associated
          Gene Aliases:
          BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
          Chromosome Location:
          Chr.17: 43044295 - 43125483 on Build GRCh38
          UniGene ID:
          Hs.194143
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536341

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BRCA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007294.3 NP_009225.1 9 844
          NM_007297.3 NP_009228.2 8 752
          NM_007298.3 NP_009229.2 8 631
          NM_007299.3 NP_009230.2 9 806
          NM_007300.3 NP_009231.2 9 844
          NR_027676.1 9 748
          GenBank mRNA AF005068.1 - 9 713
          AK307553.1 - 8 647
          AK308084.1 - 9 773
          AK316200.1 - 8 703
          AY354539.2 - 8 676
          AY751490.1 - 8 612
          BC030969.1 - 9 731
          BC072418.1 - 9 806
          BC085615.1 - 9 711
          BC106745.1 - 9 661
          BC114511.1 - 10 777
          BC115037.1 - 9 749 7961446
          JN686490.1 - 8 612
          U14680.1 - 9 731
          U64805.1 - 8 612

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          DNA double-strand break processing
          DNA synthesis involved in DNA repair
          strand displacement
          DNA replication
          postreplication repair
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase III promoter
          fatty acid biosynthetic process
          apoptotic process
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          chromosome segregation
          centrosome cycle
          brain development
          response to nutrient
          intrinsic apoptotic signaling pathway in response to DNA damage
          dosage compensation by inactivation of X chromosome
          response to ionizing radiation
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          protein ubiquitination
          protein sumoylation
          androgen receptor signaling pathway
          chromosome breakage
          positive regulation of protein ubiquitination
          G2 DNA damage checkpoint
          response to estradiol
          negative regulation of intracellular estrogen receptor signaling pathway
          positive regulation of protein import into nucleus, translocation
          positive regulation of histone acetylation
          negative regulation of histone acetylation
          regulation of cell proliferation
          regulation of apoptotic process
          chordate embryonic development
          response to estrogen
          regulation of DNA methylation
          negative regulation of fatty acid biosynthetic process
          positive regulation of DNA repair
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of centriole replication
          positive regulation of histone H3-K4 methylation
          negative regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          positive regulation of histone H3-K9 methylation
          protein autoubiquitination
          positive regulation of histone H4-K20 methylation
          positive regulation of cell cycle arrest
          cellular response to tumor necrosis factor
          cellular response to indole-3-methanol
          protein K6-linked ubiquitination
          regulation of signal transduction by p53 class mediator
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of reactive oxygen species metabolic process
          positive regulation of histone H3-K9 acetylation
          positive regulation of histone H4-K16 acetylation
          DNA binding
          chromatin binding
          damaged DNA binding
          transcription coactivator activity
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          tubulin binding
          ligase activity
          enzyme binding
          ubiquitin protein ligase binding
          transcription regulatory region DNA binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dcnqv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0