Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTM1 SIRNA Assays ›
          Gene Symbol:
          MTM1
          Gene Name:
          myotubularin 1
          Gene Aliases:
          CNM, MTMX, XLMTM
          Chromosome Location:
          Chr.X: 150562658 - 150673143 on Build GRCh38
          UniGene ID:
          Hs.655056
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536346

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000252.2 NP_000243.1 9 773
          XM_005274687.2 XP_005274744.1 9 1070
          XM_011531171.1 XP_011529473.1 10 821
          XM_011531172.1 XP_011529474.1 10 1115
          XM_011531173.2 XP_011529475.1 9 773
          XM_017029547.1 XP_016885036.1 9 752
          XM_017029548.1 XP_016885037.1 10 773
          XM_017029549.1 XP_016885038.1 9 749
          XM_017029550.1 XP_016885039.1 8 665
          XM_017029551.1 XP_016885040.1 4 4965
          GenBank mRNA AK291710.1 - 9 761
          AK297021.1 - 8 616
          AK297057.1 - 7 543
          AK304036.1 - 8 654
          AL832715.1 - 9 569
          BC030779.1 - 9 732 4824189
          U46024.1 - 9 751

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          phosphatidylinositol biosynthetic process
          endosome to lysosome transport
          protein transport
          negative regulation of TOR signaling
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          peptidyl-tyrosine dephosphorylation
          regulation of vacuole organization
          intermediate filament organization
          muscle cell cellular homeostasis
          phosphatidylinositol dephosphorylation
          mitochondrion distribution
          positive regulation of skeletal muscle tissue growth
          negative regulation of protein kinase B signaling
          mitochondrion morphogenesis
          negative regulation of autophagosome assembly
          phosphatidylinositol-3-phosphatase activity
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          intermediate filament binding
          phosphatidylinositol binding
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0