Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BMPR2 SIRNA Assays ›
          Gene Symbol:
          BMPR2
          Gene Name:
          bone morphogenetic protein receptor type 2
          Gene Aliases:
          BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
          Chromosome Location:
          Chr.2: 202376310 - 202567751 on Build GRCh38
          UniGene ID:
          Hs.471119
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536354

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BMPR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204.6 NP_001195.2 6 1932
          XM_011511687.1 XP_011509989.1 6 1949
          GenBank mRNA AK292430.1 - 6 1196
          BC052985.2 - 6 1358 6149698
          D50516.1 - 6 1192
          U20165.1 - 6 820
          Z48923.1 - 6 1165

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          maternal placenta development
          endothelial cell proliferation
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          blood vessel remodeling
          chondrocyte development
          negative regulation of systemic arterial blood pressure
          transcription from RNA polymerase II promoter
          transmembrane receptor protein serine/threonine kinase signaling pathway
          brain development
          cellular response to starvation
          anterior/posterior pattern specification
          positive regulation of endothelial cell migration
          positive regulation of epithelial cell migration
          positive regulation of pathway-restricted SMAD protein phosphorylation
          regulation of lung blood pressure
          signal transduction by protein phosphorylation
          proteoglycan biosynthetic process
          negative regulation of cell growth
          positive regulation of bone mineralization
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          activin receptor signaling pathway
          regulation of cell proliferation
          positive regulation of osteoblast differentiation
          negative regulation of vasoconstriction
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          lung alveolus development
          positive regulation of axon extension involved in axon guidance
          limb development
          endochondral bone morphogenesis
          lymphatic endothelial cell differentiation
          artery development
          venous blood vessel development
          positive regulation of cartilage development
          retina vasculature development in camera-type eye
          cellular response to BMP stimulus
          endothelial cell apoptotic process
          negative regulation of chondrocyte proliferation
          negative regulation of DNA biosynthetic process
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          activin receptor activity, type II
          growth factor binding
          BMP binding
          metal ion binding
          BMP receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2gfbh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0