Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012994.1 | NP_001013012.1 | 1, 2 | 297 | ||
XM_011518691.2 | XP_011516993.1 | 1, 2 | 334 | |||
GenBank mRNA | BG699098.1 | - | 1, 2 | 318 | ||
BX367137.2 | - | 1, 2 | 296 |