Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE3A SIRNA Assays ›
          Gene Symbol:
          UBE3A
          Gene Name:
          ubiquitin protein ligase E3A
          Gene Aliases:
          ANCR, AS, E6-AP, EPVE6AP, HPVE6A
          Chromosome Location:
          Chr.15: 25337234 - 25439086 on Build GRCh38
          UniGene ID:
          Hs.598862
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536476

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000462.3 NP_000453.2 7 2261
          NM_130838.1 NP_570853.1 3 1476
          NM_130839.2 NP_570854.1 6 2196
          XM_005268267.4 XP_005268324.1 8 2563
          XM_005268268.4 XP_005268325.1 7 2450
          XM_005268269.4 XP_005268326.1 8 2294
          XM_005268270.4 XP_005268327.1 7 2251
          XM_005268271.4 XP_005268328.1 4 2070
          XM_006720675.3 XP_006720738.1 8 2515
          XM_006720676.3 XP_006720739.1 5 1760
          XM_011521994.2 XP_011520296.1 8 2452
          XM_011521995.2 XP_011520297.1 8 2126
          XM_017022544.1 XP_016878033.1 9 2488
          XM_017022545.1 XP_016878034.1 8 2447
          XM_017022546.1 XP_016878035.1 8 2355
          XM_017022547.1 XP_016878036.1 7 2387
          XM_017022548.1 XP_016878037.1 7 2382
          XM_017022549.1 XP_016878038.1 7 2189
          XM_017022550.1 XP_016878039.1 7 2341
          XM_017022551.1 XP_016878040.1 8 2392
          XM_017022552.1 XP_016878041.1 8 2082
          XM_017022553.1 XP_016878042.1 8 2397
          XM_017022554.1 XP_016878043.1 4 2401
          XM_017022555.1 XP_016878044.1 6 2170
          XM_017022556.1 XP_016878045.1 7 2319
          XM_017022557.1 XP_016878046.1 6 2254
          XM_017022558.1 XP_016878047.1 7 2450
          XM_017022559.1 XP_016878048.1 4 2070
          GenBank mRNA AB194232.1 - 5 1572
          AF273050.1 - 4 1531
          AK130491.1 - 2 1205
          AK291405.1 - 4 1622
          AK292514.1 - 6 1806
          BC002582.2 - 6 2149 3160225
          BC009271.1 - 1 753 3353595
          L07557.1 - 3 1498
          U84404.1 - 8 2018
          X98031.1 - 4 1501
          X98032.1 - 2 1432
          X98033.1 - 4 1492

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191837, XM_008759399, XM_006229313, XM_006229310, XM_008759400, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011668, NM_173010, NM_001033962, AK083795, AK029133, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          proteolysis
          ubiquitin-dependent protein catabolic process
          brain development
          positive regulation of phosphatidylinositol 3-kinase signaling
          viral process
          androgen receptor signaling pathway
          positive regulation of protein ubiquitination
          sperm entry
          regulation of circadian rhythm
          positive regulation of transcription from RNA polymerase II promoter
          protein autoubiquitination
          prostate gland growth
          protein K48-linked ubiquitination
          regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          transcription coactivator activity
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0