Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KRIT1 SIRNA Assays ›
          Gene Symbol:
          KRIT1
          Gene Name:
          KRIT1, ankyrin repeat containing
          Gene Aliases:
          CAM, CCM1
          Chromosome Location:
          Chr.7: 92198969 - 92246128 on Build GRCh38
          UniGene ID:
          Hs.531987
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536492

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KRIT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013406.1 NP_001013424.1 4, 5 667
          NM_004912.3 NP_004903.2 5, 6 1162
          NM_194454.1 NP_919436.1 5, 6 815
          NM_194455.1 NP_919437.1 5, 6 1289
          NM_194456.1 NP_919438.1 6, 7 1054
          XM_005250660.3 XP_005250717.1 5, 6 782
          XM_005250662.3 XP_005250719.1 5, 6 761
          XM_005250665.3 XP_005250722.1 5, 6 689
          XM_005250666.3 XP_005250723.1 6, 7 865
          XM_005250667.2 XP_005250724.1 4, 5 573
          XM_005250668.3 XP_005250725.1 4, 5 635
          XM_005250669.3 XP_005250726.1 4, 5 613
          XM_006716161.3 XP_006716224.1 5, 6 913
          XM_006716162.3 XP_006716225.1 5, 6 730
          XM_006716163.3 XP_006716226.1 4, 5 580
          XM_011516651.2 XP_011514953.1 6, 7 896
          XM_011516653.2 XP_011514955.1 4, 5 650
          XM_011516654.2 XP_011514956.1 6, 7 914
          XM_011516655.2 XP_011514957.1 4, 5 568
          XM_011516656.2 XP_011514958.1 4, 5 537
          XM_011516657.2 XP_011514959.1 4, 5 457
          XM_011516658.2 XP_011514960.1 4, 5 469
          XM_011516659.2 XP_011514961.1 4, 5 491
          XM_011516660.2 XP_011514962.1 3, 4 373
          XM_011516661.2 XP_011514963.1 3, 4 451
          XM_017012755.1 XP_016868244.1 4, 5 540
          XM_017012756.1 XP_016868245.1 4, 5 502
          XM_017012757.1 XP_016868246.1 4, 5 534
          XM_017012758.1 XP_016868247.1 5, 6 1140
          XM_017012759.1 XP_016868248.1 4, 5 992
          GenBank mRNA AF296765.2 - 5, 6 1059
          AF310133.1 - 5, 6 754
          AF388384.1 - 5, 6 1289
          AJ294850.1 - 6, 7 1013
          AK055305.1 - 4, 5 667
          AK056537.1 - 4, 5 597
          AY380057.1 - 5, 6 754
          AY380058.1 - 5, 6 754
          AY380059.1 - 5, 6 754
          AY380060.1 - 5, 6 754
          AY993944.1 - 2, 3 270
          BC094684.1 - 2, 3 280 6174749
          BX648659.1 - 1, 2 135

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          negative regulation of endothelial cell proliferation
          small GTPase mediated signal transduction
          negative regulation of endothelial cell migration
          negative regulation of angiogenesis
          positive regulation of protein binding
          cell redox homeostasis
          regulation of catalytic activity
          regulation of establishment of cell polarity
          negative regulation of endothelial cell apoptotic process
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          microtubule binding
          GTPase regulator activity
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0