Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GBA SIRNA Assays ›
          Gene Symbol:
          GBA GBAP1
          Gene Name:
          glucosylceramidase beta
          glucosylceramidase beta pseudogene 1
          Gene Aliases:
          GBA1, GCB, GLUC
          GBAP
          Chromosome Location:
          Chr.1: 155234448 - 155244862 on Build GRCh38
          Chromosome Location:
          Chr.1: 155213825 - 155227534 on Build GRCh38
          UniGene ID:
          Hs.282997
          Hs.719930
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s536661

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GBA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000157.3 NP_000148.2 9 1459
          NM_001005741.2 NP_001005741.1 10 1718
          NM_001005742.2 NP_001005742.1 10 1699
          NM_001171811.1 NP_001165282.1 8 1535
          NM_001171812.1 NP_001165283.1 8 1309
          GenBank mRNA AK291911.1 - 10 1697
          AK298900.1 - 8 1309
          AK300186.1 - 8 1379
          AK300829.1 - 8 1372
          AK300876.1 - 10 1699
          AK301374.1 - 7 1068
          AK302000.1 - 8 1284
          AK312502.1 - 10 1515
          BC003356.1 - 9 1398 2899915
          BX648487.1 - 10 1527
          D13286.1 - 9 1398
          K02920.1 - 8 1278
          KJ690771.1 - 9 1342
          M16328.1 - 9 1728
          M19285.1 - 9 1415
          Gene Symbol: GBAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA KJ690774.1 - 1093
          KJ690776.1 - 983

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glucosylceramide catabolic process
          glycosphingolipid metabolic process
          cellular response to starvation
          response to pH
          positive regulation of macroautophagy
          regulation of macroautophagy
          termination of signal transduction
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of interleukin-6 production
          regulation of water loss via skin
          response to testosterone
          positive regulation of protein dephosphorylation
          negative regulation of MAP kinase activity
          skin morphogenesis
          response to estrogen
          sphingosine biosynthetic process
          ceramide biosynthetic process
          negative regulation of inflammatory response
          response to glucocorticoid
          cellular response to tumor necrosis factor
          response to thyroid hormone
          positive regulation of proteolysis involved in cellular protein catabolic process
          glucosylceramidase activity
          receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0