siRNA Details
Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI701640.1 | - | 1 | 39 | ||
AW069047.1 | - | 1 | 61 | |||
BQ574961.1 | - | 1 | 77 |

Gene Symbol: ZNF503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI701640.1 | - | 1 | 39 | ||
AW069047.1 | - | 1 | 61 | |||
BQ574961.1 | - | 1 | 77 |