Search
Search

RNASEK| Gene Symbol: RNASEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004333.4 | NP_001004333.2 | 1, 2 | 343 | ||
| GenBank mRNA | AK289930.1 | - | 1, 2 | 126 | ||
| AM746459.1 | - | 1, 2 | 110 | |||
| BC051802.1 | - | 1, 2 | 114 | 4346419 | ||
| BC062705.1 | - | 1, 2 | 118 | 4273688 | ||
| BM835723.1 | - | 1, 2 | 343 | |||
| CB160051.1 | - | 1, 2 | 281 | |||