Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other XRCC6 SIRNA Assays ›
          Gene Symbol:
          XRCC6 XRCC6P5
          Gene Name:
          X-ray repair cross complementing 6
          X-ray repair cross complementing 6 pseudogene 5
          Gene Aliases:
          CTC75, CTCBF, G22P1, KU70, ML8, TLAA
          -
          Chromosome Location:
          Chr.22: 41621163 - 41664048 on Build GRCh38
          Chromosome Location:
          Chr.X: 99461602 - 99939843 on Build GRCh38
          UniGene ID:
          Hs.292493
          Hs.706599
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s5456

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: XRCC6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288976.1 NP_001275905.1 6, 7 865
          NM_001288977.1 NP_001275906.1 6, 7 856
          NM_001288978.1 NP_001275907.1 5, 6 866
          NM_001469.4 NP_001460.1 6, 7 979
          GenBank mRNA AK055786.1 - 6, 7 724
          AK293439.1 - 5, 6 713
          AK304580.1 - 5, 6 722
          AK315618.1 - 6, 7 873
          BC001583.1 - 3, 4 529
          BC008343.1 - 5, 6 1436 3509801
          BC010034.2 - 6, 7 820 3536083
          BC012154.2 - 5, 6 797 4561541
          BC018259.2 - 6, 7 819 3448510
          BC072449.1 - 5, 6 803 6167735
          BQ217843.1 - 2, 3 462
          BU902323.1 - 6, 7 870
          CR542219.1 - 5, 6 781
          J04607.1 - 5, 6 798
          J04611.1 - 6, 7 814
          M32865.1 - 5, 6 805
          S38729.1 - 5, 6 805
          Gene Symbol: XRCC6P5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024608.1 8 958
          GenBank mRNA EU224311.1 - 4 976

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA ligation
          double-strand break repair via nonhomologous end joining
          DNA recombination
          transcription, DNA-templated
          positive regulation of type I interferon production
          DNA duplex unwinding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of smooth muscle cell proliferation
          protein heterotetramerization
          cellular response to gamma radiation
          cellular response to X-ray
          establishment of integrated proviral latency
          double-strand break repair via classical nonhomologous end joining
          DNA binding
          damaged DNA binding
          double-stranded DNA binding
          double-stranded telomeric DNA binding
          ATP-dependent DNA helicase activity
          protein binding
          ATP binding
          protein C-terminus binding
          transcription regulatory region DNA binding
          poly(A) RNA binding
          macromolecular complex binding
          5'-deoxyribose-5-phosphate lyase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0