Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003702.2 | NP_001003702.2 | 2 | 1820 | ||
GenBank mRNA | AK125680.1 | - | 2 | 1765 | ||
BC136868.1 | - | 2 | 1679 | 9020859 | ||
BC136878.1 | - | 2 | 1679 | 9020869 | ||
DQ786206.1 | - | 1 | 438 | |||
EF560743.1 | - | 2 | 1699 |