Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CT45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017417.2 | NP_001017417.1 | 3 | 612 | ||
XM_005278141.3 | XP_005278198.1 | 3 | 461 | |||
XM_011531352.1 | XP_011529654.1 | 2 | 382 | |||
GenBank mRNA | AY743709.1 | - | 3 | 613 | ||
BC140913.1 | - | 3 | 512 | 9056725 |
Gene Symbol: CT45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017435.1 | NP_001017435.1 | 3 | 613 | ||
XM_006724758.3 | XP_006724821.1 | 3 | 530 | |||
XM_011531340.2 | XP_011529642.1 | 3 | 489 | |||
GenBank mRNA | AY743711.1 | - | 3 | 613 |
Gene Symbol: CT45A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007551.4 | NP_001007552.2 | 3 | 613 | ||
NM_001172288.2 | NP_001165759.2 | 3 | 630 | |||
XM_011531341.2 | XP_011529643.1 | 3 | 495 | |||
GenBank mRNA | BC028711.2 | - | 3 | 613 | 4828274 | |
BC140735.1 | - | 3 | 520 | 9021603 | ||
BC140926.1 | - | 3 | 520 | 9056738 | ||
BC144437.1 | - | 3 | 520 | 9052959 |
Gene Symbol: CT45A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017438.2 | NP_001017438.2 | 3 | 613 | ||
XM_005262425.4 | XP_005262482.2 | 3 | 533 | |||
XM_011531351.1 | XP_011529653.1 | 3 | 464 |
Gene Symbol: CT45A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291543.1 | NP_001278472.1 | 3 | 429 | ||
XM_003960082.4 | XP_003960131.1 | 3 | 516 | |||
XM_011531229.2 | XP_011529531.1 | 3 | 495 | |||
GenBank mRNA | BU540102.1 | - | 3 | 432 |