Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CKMT1A SIRNA Assays ›
          Gene Symbol:
          CKMT1A CKMT1B
          Gene Name:
          creatine kinase, mitochondrial 1A
          creatine kinase, mitochondrial 1B
          Gene Aliases:
          CKMT1, U-MtCK, mia-CK
          CKMT, CKMT1, UMTCK
          Chromosome Location:
          Chr.15: 43692644 - 43699222 on Build GRCh38
          Chromosome Location:
          Chr.15: 43592857 - 43599406 on Build GRCh38
          UniGene ID:
          Hs.654988
          Hs.741420
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s54725

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CKMT1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001015001.2 NP_001015001.1 7 1286
          NM_001321926.1 NP_001308855.1 6 1154
          NM_001321927.1 NP_001308856.1 7 1379
          NM_001321928.1 NP_001308857.1 7 1140
          NM_001321929.1 NP_001308858.1 6 1138
          NR_135856.1 6 1385
          XM_005254498.3 XP_005254555.1 6 861
          XM_017022369.1 XP_016877858.1 7 1211
          XM_017022370.1 XP_016877859.1 7 1136
          GenBank mRNA AK223365.1 - 6 966
          AK295776.1 - 6 1030
          AK316052.1 - 7 786
          AK316124.1 - 7 1237
          AK316319.1 - 7 1140
          BC001926.1 - 6 971 2820642
          BC006467.1 - 6 897 3535486
          BT006628.1 - 6 794
          Gene Symbol: CKMT1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020990.4 NP_066270.1 7 1383
          NR_135748.1 6 1432
          NR_135749.1 6 1412
          NR_135750.1 7 1383
          NR_135751.1 6 1272
          NR_135752.1 5 1039
          XM_005254150.3 XP_005254207.1 6 861
          XM_011521194.1 XP_011519496.1 7 1471
          XM_011521195.2 XP_011519497.1 7 1210
          XM_011521196.1 XP_011519498.1 7 1136
          XM_011521197.2 XP_011519499.1 6 970
          XM_011521198.1 XP_011519500.1 7 1655
          XM_011521199.2 XP_011519501.2 7 1427
          XM_017021902.1 XP_016877391.1 7 1611
          GenBank mRNA AK094322.1 - 7 961
          AK293685.1 - 6 653
          AK293790.1 - 6 1239
          AK293939.1 - 6 1036
          AK293961.1 - 5 821
          AK294063.1 - 6 722
          AK294735.1 - 7 854
          AK294908.1 - 6 1271
          AK296271.1 - 5 472
          AK296472.1 - 6 874
          AK308770.1 - 5 744
          BC108652.1 - 6 906 6190035
          BC121001.1 - 6 970 40116549
          BC121002.1 - 6 970 40116552

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          amino acid kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          creatine metabolic process
          phosphorylation
          creatine kinase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0