Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100916.1 | NP_001094386.1 | 3 | 799 | ||
XM_017013726.1 | XP_016869215.1 | 4 | 1402 | |||
GenBank mRNA | AF359269.1 | - | 1 | 419 | ||
EU518495.2 | - | 3 | 809 |