Search
Search

GALK1| Gene Symbol: GALK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000154.1 | NP_000145.1 | 4 | 627 | ||
| GenBank mRNA | AK130064.1 | - | 917 | |||
| AK303748.1 | - | 3 | 771 | |||
| AK310203.1 | - | 4 | 877 | |||
| AK314890.1 | - | 4 | 587 | |||
| AK316181.1 | - | 3 | 755 | |||
| BC001166.1 | - | 4 | 608 | 3501788 | ||
| BT007005.1 | - | 4 | 564 | |||
| EU794610.1 | - | 4 | 592 | |||
| U26401.1 | - | 4 | 592 | |||