Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080529.2 | NP_001073998.2 | 5 | 913 | ||
XM_017012522.1 | XP_016868011.1 | 4 | 921 | |||
GenBank mRNA | AK094250.1 | - | 1 | 70 |