Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTBP2 SIRNA Assays ›
Gene Symbol:
CTBP2
Gene Name:
C-terminal binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 124987849 - 125161061 on Build GRCh38
UniGene ID:
Hs.501345
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s56075
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083914.2
NP_001077383.1
7
1204
NM_001290214.2
NP_001277143.1
7
1189
NM_001290215.2
NP_001277144.1
7
1453
NM_001321012.1
NP_001307941.1
7
1148
NM_001321013.1
NP_001307942.1
6
1349
NM_001321014.1
NP_001307943.1
7
1450
NM_001329.3
NP_001320.1
7
1192
NM_022802.2
NP_073713.2
5
2509
XM_005269561.2
XP_005269618.1
8
1375
XM_005269564.2
XP_005269621.1
8
1583
XM_005269567.2
XP_005269624.1
8
1582
XM_005269568.4
XP_005269625.1
8
1310
XM_005269569.2
XP_005269626.1
7
1271
XM_005269571.2
XP_005269628.1
6
1030
XM_005269572.3
XP_005269629.1
6
1078
XM_006717642.2
XP_006717705.1
9
1504
XM_011539349.2
XP_011537651.1
5
1286
XM_011539351.1
XP_011537653.1
10
1605
XM_011539353.1
XP_011537655.1
9
1476
XM_011539354.1
XP_011537656.1
8
1400
XM_011539355.1
XP_011537657.1
7
1478
XM_011539357.1
XP_011537659.1
7
1216
XM_011539358.1
XP_011537660.1
6
1294
XM_017015756.1
XP_016871245.1
8
1246
XM_017015757.1
XP_016871246.1
7
2972
GenBank mRNA
AF016507.1
-
7
1109
AF222711.1
-
5
2526
AK290390.1
-
7
1053
AL833398.1
-
7
1164
BC002486.2
-
7
1165
BC037900.2
-
5
1173
5276086
BC047018.1
-
7
1169
BC052276.1
-
7
1146
BC072020.1
-
7
1126
BT007012.1
-
5
764
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cell proliferation
viral genome replication
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
white fat cell differentiation
oxidation-reduction process
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
retinoic acid receptor binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0