Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57313
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198834.3
NP_001185763.3
24
3691
NM_014644.5
NP_055459.5
24
3691
XM_005272981.3
XP_005273038.2
20
4318
XM_005277442.4
XP_005277499.2
20
4318
XM_005277443.3
XP_005277500.1
20
4318
XM_006711642.2
XP_006711705.1
20
4318
XM_006711643.2
XP_006711706.1
20
4318
XM_006711644.2
XP_006711707.1
20
4318
XM_006711646.1
XP_006711709.1
27
3980
XM_006711650.3
XP_006711713.1
25
3465
XM_006711651.1
XP_006711714.1
24
3583
XM_006711652.1
XP_006711715.1
24
3583
XM_011510172.1
XP_011508474.1
27
4138
XM_011510173.1
XP_011508475.1
27
4138
XM_011510174.1
XP_011508476.1
20
4318
XM_011510175.1
XP_011508477.1
27
4138
XM_011510179.1
XP_011508481.1
24
3583
XM_017002875.1
XP_016858364.1
20
4318
XM_017002876.1
XP_016858365.1
20
4318
XM_017002877.1
XP_016858366.1
20
4318
XM_017002878.1
XP_016858367.1
27
4138
XM_017002879.1
XP_016858368.1
20
4318
XM_017002880.1
XP_016858369.1
27
4138
XM_017002881.1
XP_016858370.1
20
4318
XM_017002882.1
XP_016858371.1
20
4318
XM_017002883.1
XP_016858372.1
20
4318
XM_017002884.1
XP_016858373.1
20
4318
XM_017002887.1
XP_016858376.1
28
4415
XM_017002888.1
XP_016858377.1
28
4388
XM_017002889.1
XP_016858378.1
28
4060
XM_017002890.1
XP_016858379.1
20
4318
XM_017002891.1
XP_016858380.1
26
4219
XM_017002892.1
XP_016858381.1
26
3850
XM_017002893.1
XP_016858382.1
27
4338
XM_017002894.1
XP_016858383.1
27
3983
XM_017002895.1
XP_016858384.1
24
3301
XM_017002896.1
XP_016858385.1
20
4318
XM_017002897.1
XP_016858386.1
20
4318
XM_017002898.1
XP_016858387.1
27
3980
GenBank mRNA
AL833273.1
-
24
3579
BX647660.1
-
24
3619
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
CR749273.1
-
5
823
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline