Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57383
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002811.2
NP_001002811.2
4
2057
NM_001002812.2
NP_001002812.2
8
1213
NM_001198832.2
NP_001185761.2
11
1495
NM_001198834.3
NP_001185763.3
8
1213
NM_014644.5
NP_055459.5
8
1213
XM_005272981.3
XP_005273038.2
4
1840
XM_005277442.4
XP_005277499.2
4
1840
XM_005277443.3
XP_005277500.1
4
1840
XM_006711642.2
XP_006711705.1
4
1840
XM_006711643.2
XP_006711706.1
4
1840
XM_006711644.2
XP_006711707.1
4
1840
XM_006711646.1
XP_006711709.1
11
1502
XM_006711648.2
XP_006711711.1
4
1840
XM_006711650.3
XP_006711713.1
9
987
XM_006711651.1
XP_006711714.1
8
1105
XM_006711652.1
XP_006711715.1
8
1105
XM_011510172.1
XP_011508474.1
11
1660
XM_011510173.1
XP_011508475.1
11
1660
XM_011510174.1
XP_011508476.1
4
1840
XM_011510175.1
XP_011508477.1
11
1660
XM_011510176.1
XP_011508478.1
11
1660
XM_011510179.1
XP_011508481.1
8
1105
XM_017002875.1
XP_016858364.1
4
1840
XM_017002876.1
XP_016858365.1
4
1840
XM_017002877.1
XP_016858366.1
4
1840
XM_017002878.1
XP_016858367.1
11
1660
XM_017002879.1
XP_016858368.1
4
1840
XM_017002880.1
XP_016858369.1
11
1660
XM_017002881.1
XP_016858370.1
4
1840
XM_017002882.1
XP_016858371.1
4
1840
XM_017002883.1
XP_016858372.1
4
1840
XM_017002884.1
XP_016858373.1
4
1840
XM_017002885.1
XP_016858374.1
4
1840
XM_017002886.1
XP_016858375.1
4
1840
XM_017002887.1
XP_016858376.1
12
1937
XM_017002888.1
XP_016858377.1
12
1910
XM_017002889.1
XP_016858378.1
12
1582
XM_017002890.1
XP_016858379.1
4
1840
XM_017002891.1
XP_016858380.1
10
1741
XM_017002892.1
XP_016858381.1
10
1372
XM_017002893.1
XP_016858382.1
11
1860
XM_017002894.1
XP_016858383.1
11
1505
XM_017002895.1
XP_016858384.1
8
823
XM_017002896.1
XP_016858385.1
4
1840
XM_017002897.1
XP_016858386.1
4
1840
XM_017002898.1
XP_016858387.1
11
1502
XM_017002899.1
XP_016858388.1
11
1502
XM_017002900.1
XP_016858389.1
4
1840
XM_017002901.1
XP_016858390.1
4
1840
XM_017002902.1
XP_016858391.1
11
1502
XM_017002903.1
XP_016858392.1
11
1502
GenBank mRNA
AB007923.2
-
11
1374
AB007946.1
-
4
2057
AL831815.2
-
8
1056
AL832024.2
-
8
1021
AL833273.1
-
8
1101
BC004860.2
-
4
1724
BC078660.1
-
4
1563
BC132717.1
-
4
1660
BC152439.1
-
4
2057
BX647660.1
-
8
1141
HQ333476.1
-
4
1470
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003064.1
XP_016858553.1
11
1656
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5x9lz:80/100.66.76.20:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0