Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36B SIRNA Assays ›
Gene Symbol:
ANKRD36B
ANKRD36
ANKRD36C
Gene Name:
ankyrin repeat domain 36B
ankyrin repeat domain 36
ankyrin repeat domain 36C
Gene Aliases:
KIAA1641
UNQ2430
-
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 95849548 - 95992320 on Build GRCh38
UniGene ID:
Hs.532921
Hs.541894
Hs.646318
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
siRNA ID
s57459
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025190.3
NP_079466.3
3
683
XM_005263995.4
XP_005264052.1
3
963
XM_006712661.1
XP_006712724.1
3
963
XM_011511555.1
XP_011509857.1
3
963
XM_011511556.1
XP_011509858.1
3
963
XM_011511557.2
XP_011509859.1
3
963
XM_011511558.1
XP_011509860.1
3
963
XM_017004578.1
XP_016860067.1
3
963
XM_017004579.1
XP_016860068.1
3
963
XM_017004580.1
XP_016860069.1
3
963
XM_017004581.1
XP_016860070.1
3
963
XM_017004582.1
XP_016860071.1
3
963
XM_017004583.1
XP_016860072.1
3
963
XM_017004584.1
XP_016860073.1
3
963
XM_017004585.1
XP_016860074.1
3
963
XM_017004587.1
XP_016860076.1
3
963
XM_017004588.1
XP_016860077.1
3
963
XM_017004589.1
XP_016860078.1
3
963
XM_017004590.1
XP_016860079.1
3
963
XM_017004592.1
XP_016860081.1
3
963
XM_017004593.1
XP_016860082.1
3
963
XM_017004594.1
XP_016860083.1
3
963
XM_017004595.1
XP_016860084.1
3
963
GenBank mRNA
AK074570.1
-
3
584
BC071771.1
-
3
509
BM511319.1
-
3
275
DR001190.1
-
3
572
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
3
683
XM_006712514.3
XP_006712577.1
3
993
XM_011511130.1
XP_011509432.1
3
993
XM_011511131.1
XP_011509433.1
3
993
XM_017004009.1
XP_016859498.1
3
993
XM_017004010.1
XP_016859499.1
3
993
XM_017004011.1
XP_016859500.1
3
993
XM_017004012.1
XP_016859501.1
3
993
XM_017004013.1
XP_016859502.1
3
993
XM_017004014.1
XP_016859503.1
3
993
XM_017004015.1
XP_016859504.1
3
993
XM_017004016.1
XP_016859505.1
3
993
XM_017004017.1
XP_016859506.1
3
993
XM_017004018.1
XP_016859507.1
3
993
XM_017004019.1
XP_016859508.1
3
993
XM_017004020.1
XP_016859509.1
3
993
XM_017004021.1
XP_016859510.1
3
993
XM_017004022.1
XP_016859511.1
3
993
XM_017004023.1
XP_016859512.1
3
993
XM_017004024.1
XP_016859513.1
3
993
XM_017004025.1
XP_016859514.1
3
993
XM_017004026.1
XP_016859515.1
3
993
XM_017004027.1
XP_016859516.1
3
993
XM_017004028.1
XP_016859517.1
3
993
XM_017004029.1
XP_016859518.1
3
993
XM_017004030.1
XP_016859519.1
3
993
XM_017004031.1
XP_016859520.1
3
993
XM_017004032.1
XP_016859521.1
3
993
XM_017004033.1
XP_016859522.1
3
993
XM_017004034.1
XP_016859523.1
3
993
XM_017004035.1
XP_016859524.1
3
993
XM_017004036.1
XP_016859525.1
3
993
XM_017004037.1
XP_016859526.1
3
993
XM_017004038.1
XP_016859527.1
3
993
XM_017004039.1
XP_016859528.1
3
993
XM_017004040.1
XP_016859529.1
3
993
XM_017004041.1
XP_016859530.1
3
993
XM_017004042.1
XP_016859531.1
3
993
XM_017004043.1
XP_016859532.1
3
993
XM_017004044.1
XP_016859533.1
3
993
XM_017004045.1
XP_016859534.1
3
993
XM_017004046.1
XP_016859535.1
3
993
XM_017004047.1
XP_016859536.1
3
993
XM_017004048.1
XP_016859537.1
3
993
XM_017004049.1
XP_016859538.1
3
993
GenBank mRNA
AK304740.1
-
3
623
AK307629.1
-
3
590
AK316541.1
-
3
623
AY358571.1
-
3
682
BC046186.1
-
3
562
BC128262.1
-
3
522
BC128517.1
-
3
522
40118468
BX647769.1
-
3
628
Gene Symbol: ANKRD36C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001310154.1
NP_001297083.1
3
1051
GenBank mRNA
AK302433.1
-
3
525
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
ion channel inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0