Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36 SIRNA Assays ›
Gene Symbol:
ANKRD36
ANKRD36B
Gene Name:
ankyrin repeat domain 36
ankyrin repeat domain 36B
Gene Aliases:
UNQ2430
KIAA1641
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
UniGene ID:
Hs.541894
Hs.532921
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57460
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
3
633
XM_006712514.3
XP_006712577.1
3
943
XM_011511130.1
XP_011509432.1
3
943
XM_011511131.1
XP_011509433.1
3
943
XM_017004009.1
XP_016859498.1
3
943
XM_017004010.1
XP_016859499.1
3
943
XM_017004011.1
XP_016859500.1
3
943
XM_017004012.1
XP_016859501.1
3
943
XM_017004013.1
XP_016859502.1
3
943
XM_017004014.1
XP_016859503.1
3
943
XM_017004015.1
XP_016859504.1
3
943
XM_017004016.1
XP_016859505.1
3
943
XM_017004017.1
XP_016859506.1
3
943
XM_017004018.1
XP_016859507.1
3
943
XM_017004019.1
XP_016859508.1
3
943
XM_017004020.1
XP_016859509.1
3
943
XM_017004021.1
XP_016859510.1
3
943
XM_017004022.1
XP_016859511.1
3
943
XM_017004023.1
XP_016859512.1
3
943
XM_017004024.1
XP_016859513.1
3
943
XM_017004025.1
XP_016859514.1
3
943
XM_017004026.1
XP_016859515.1
3
943
XM_017004027.1
XP_016859516.1
3
943
XM_017004028.1
XP_016859517.1
3
943
XM_017004029.1
XP_016859518.1
3
943
XM_017004030.1
XP_016859519.1
3
943
XM_017004031.1
XP_016859520.1
3
943
XM_017004032.1
XP_016859521.1
3
943
XM_017004033.1
XP_016859522.1
3
943
XM_017004034.1
XP_016859523.1
3
943
XM_017004035.1
XP_016859524.1
3
943
XM_017004036.1
XP_016859525.1
3
943
XM_017004037.1
XP_016859526.1
3
943
XM_017004038.1
XP_016859527.1
3
943
XM_017004039.1
XP_016859528.1
3
943
XM_017004040.1
XP_016859529.1
3
943
XM_017004041.1
XP_016859530.1
3
943
XM_017004042.1
XP_016859531.1
3
943
XM_017004043.1
XP_016859532.1
3
943
XM_017004044.1
XP_016859533.1
3
943
XM_017004045.1
XP_016859534.1
3
943
XM_017004046.1
XP_016859535.1
3
943
XM_017004047.1
XP_016859536.1
3
943
XM_017004048.1
XP_016859537.1
3
943
XM_017004049.1
XP_016859538.1
3
943
GenBank mRNA
AK304740.1
-
3
573
AK307629.1
-
3
540
AK316541.1
-
3
573
AY358571.1
-
3
632
BC046186.1
-
3
512
BC128262.1
-
3
472
BC128517.1
-
3
472
40118468
BX647769.1
-
3
578
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK074570.1
-
3
534
BM511319.1
-
3
225
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-p8qt9:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0