Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE4DIP SIRNA Assays ›
          Gene Symbol:
          PDE4DIP LOC100996724
          Gene Name:
          phosphodiesterase 4D interacting protein
          phosphodiesterase 4D interacting protein-like
          Gene Aliases:
          CMYA2, MMGL
          -
          Chromosome Location:
          Chr.1: 148808167 - 149051277 on Build GRCh38
          Chromosome Location:
          Chr.1: 120471449 - 120701470 on Build GRCh38
          UniGene ID:
          Hs.584841
          Hs.731111
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s57535

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDE4DIP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002811.2 NP_001002811.2 11 3152
          NM_001002812.2 NP_001002812.2 15 2308
          NM_001198832.2 NP_001185761.2 18 2590
          NM_001198834.3 NP_001185763.3 15 2308
          NM_014644.5 NP_055459.5 15 2308
          XM_005272981.3 XP_005273038.2 11 2935
          XM_005277442.4 XP_005277499.2 11 2935
          XM_005277443.3 XP_005277500.1 11 2935
          XM_006711642.2 XP_006711705.1 11 2935
          XM_006711643.2 XP_006711706.1 11 2935
          XM_006711644.2 XP_006711707.1 11 2935
          XM_006711646.1 XP_006711709.1 18 2597
          XM_006711648.2 XP_006711711.1 11 2935
          XM_006711650.3 XP_006711713.1 16 2082
          XM_006711651.1 XP_006711714.1 15 2200
          XM_006711652.1 XP_006711715.1 15 2200
          XM_011510172.1 XP_011508474.1 18 2755
          XM_011510173.1 XP_011508475.1 18 2755
          XM_011510174.1 XP_011508476.1 11 2935
          XM_011510175.1 XP_011508477.1 18 2755
          XM_011510176.1 XP_011508478.1 18 2755
          XM_011510179.1 XP_011508481.1 15 2200
          XM_017002875.1 XP_016858364.1 11 2935
          XM_017002876.1 XP_016858365.1 11 2935
          XM_017002877.1 XP_016858366.1 11 2935
          XM_017002878.1 XP_016858367.1 18 2755
          XM_017002879.1 XP_016858368.1 11 2935
          XM_017002880.1 XP_016858369.1 18 2755
          XM_017002881.1 XP_016858370.1 11 2935
          XM_017002882.1 XP_016858371.1 11 2935
          XM_017002883.1 XP_016858372.1 11 2935
          XM_017002884.1 XP_016858373.1 11 2935
          XM_017002885.1 XP_016858374.1 11 2935
          XM_017002886.1 XP_016858375.1 11 2935
          XM_017002887.1 XP_016858376.1 19 3032
          XM_017002888.1 XP_016858377.1 19 3005
          XM_017002889.1 XP_016858378.1 19 2677
          XM_017002890.1 XP_016858379.1 11 2935
          XM_017002891.1 XP_016858380.1 17 2836
          XM_017002892.1 XP_016858381.1 17 2467
          XM_017002893.1 XP_016858382.1 18 2955
          XM_017002894.1 XP_016858383.1 18 2600
          XM_017002895.1 XP_016858384.1 15 1918
          XM_017002896.1 XP_016858385.1 11 2935
          XM_017002897.1 XP_016858386.1 11 2935
          XM_017002898.1 XP_016858387.1 18 2597
          XM_017002899.1 XP_016858388.1 18 2597
          XM_017002900.1 XP_016858389.1 11 2935
          XM_017002901.1 XP_016858390.1 11 2935
          XM_017002902.1 XP_016858391.1 18 2597
          XM_017002903.1 XP_016858392.1 18 2597
          GenBank mRNA AB007923.2 - 18 2469
          AB007946.1 - 11 3152
          AL831815.2 - 15 2151
          AL832024.2 - 15 2116
          AL833273.1 - 15 2196
          BC078660.1 - 11 2658
          BC132717.1 - 11 2755
          BC152439.1 - 11 3152
          BU075360.1 - 4 542
          BX647660.1 - 15 2236
          HQ333476.1 - 11 2565
          Gene Symbol: LOC100996724
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017003064.1 XP_016858553.1 18 2751
          GenBank mRNA AK001619.1 - 7 881

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein complex assembly
          protein binding
          enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline