Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s57537
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002811.2
NP_001002811.2
12
3353
NM_001002812.2
NP_001002812.2
16
2509
NM_001198832.2
NP_001185761.2
19
2791
NM_001198834.3
NP_001185763.3
16
2509
NM_014644.5
NP_055459.5
16
2509
XM_005272981.3
XP_005273038.2
12
3136
XM_005277442.4
XP_005277499.2
12
3136
XM_005277443.3
XP_005277500.1
12
3136
XM_006711642.2
XP_006711705.1
12
3136
XM_006711643.2
XP_006711706.1
12
3136
XM_006711644.2
XP_006711707.1
12
3136
XM_006711646.1
XP_006711709.1
19
2798
XM_006711648.2
XP_006711711.1
12
3136
XM_006711650.3
XP_006711713.1
17
2283
XM_006711651.1
XP_006711714.1
16
2401
XM_006711652.1
XP_006711715.1
16
2401
XM_011510172.1
XP_011508474.1
19
2956
XM_011510173.1
XP_011508475.1
19
2956
XM_011510174.1
XP_011508476.1
12
3136
XM_011510175.1
XP_011508477.1
19
2956
XM_011510176.1
XP_011508478.1
19
2956
XM_011510179.1
XP_011508481.1
16
2401
XM_017002875.1
XP_016858364.1
12
3136
XM_017002876.1
XP_016858365.1
12
3136
XM_017002877.1
XP_016858366.1
12
3136
XM_017002878.1
XP_016858367.1
19
2956
XM_017002879.1
XP_016858368.1
12
3136
XM_017002880.1
XP_016858369.1
19
2956
XM_017002881.1
XP_016858370.1
12
3136
XM_017002882.1
XP_016858371.1
12
3136
XM_017002883.1
XP_016858372.1
12
3136
XM_017002884.1
XP_016858373.1
12
3136
XM_017002885.1
XP_016858374.1
12
3136
XM_017002886.1
XP_016858375.1
12
3136
XM_017002887.1
XP_016858376.1
20
3233
XM_017002888.1
XP_016858377.1
20
3206
XM_017002889.1
XP_016858378.1
20
2878
XM_017002890.1
XP_016858379.1
12
3136
XM_017002891.1
XP_016858380.1
18
3037
XM_017002892.1
XP_016858381.1
18
2668
XM_017002893.1
XP_016858382.1
19
3156
XM_017002894.1
XP_016858383.1
19
2801
XM_017002895.1
XP_016858384.1
16
2119
XM_017002896.1
XP_016858385.1
12
3136
XM_017002897.1
XP_016858386.1
12
3136
XM_017002898.1
XP_016858387.1
19
2798
XM_017002899.1
XP_016858388.1
19
2798
XM_017002900.1
XP_016858389.1
12
3136
XM_017002901.1
XP_016858390.1
12
3136
XM_017002902.1
XP_016858391.1
19
2798
XM_017002903.1
XP_016858392.1
19
2798
GenBank mRNA
AB007923.2
-
19
2670
AB007946.1
-
12
3353
AL831815.2
-
16
2352
AL832024.2
-
16
2317
AL833273.1
-
16
2397
BC078660.1
-
12
2859
BC132717.1
-
12
2956
BC152439.1
-
12
3353
BE536412.1
-
1
26
BX647660.1
-
16
2437
HQ333476.1
-
12
2766
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003064.1
XP_016858553.1
19
2952
GenBank mRNA
AK001619.1
-
8
1082
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline