Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GJA1 SIRNA Assays ›
          Gene Symbol:
          GJA1
          Gene Name:
          gap junction protein alpha 1
          Gene Aliases:
          AVSD3, CMDR, CX43, EKVP, GJAL, HLHS1, HSS, ODDD, PPKCA
          Chromosome Location:
          Chr.6: 121435577 - 121449744 on Build GRCh38
          UniGene ID:
          Hs.700699
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5759

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GJA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000165.4 NP_000156.1 2 738
          GenBank mRNA AK223289.1 - 2 669
          AK295460.1 - 3 583
          AK297402.1 - 2 430
          AK297764.1 - 3 622
          AK312324.1 - 2 680
          BC026329.1 - 2 554 4794131
          CR541660.1 - 1 466
          M65188.1 - 2 631
          X52947.1 - 2 623

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          neuron migration
          negative regulation of endothelial cell proliferation
          heart looping
          regulation of heart rate
          epithelial cell maturation
          lens development in camera-type eye
          chronic inflammatory response
          positive regulation of glomerular filtration
          endothelium development
          atrial ventricular junction remodeling
          transport
          apoptotic process
          muscle contraction
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          heart development
          adult heart development
          response to pH
          response to glucose
          vascular transport
          positive regulation of gene expression
          negative regulation of gene expression
          cell communication by chemical coupling
          cell communication by electrical coupling
          positive regulation of cell communication by chemical coupling
          ATP transport
          gap junction assembly
          negative regulation of cell growth
          regulation of bone mineralization
          positive regulation of insulin secretion
          ion transmembrane transport
          response to fluid shear stress
          embryonic digit morphogenesis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          skeletal muscle tissue regeneration
          response to peptide hormone
          positive regulation of osteoblast differentiation
          positive regulation of protein catabolic process
          positive regulation of striated muscle tissue development
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          regulation of bone remodeling
          blood vessel morphogenesis
          neuron projection morphogenesis
          protein oligomerization
          regulation of calcium ion transport
          negative regulation of cardiac muscle cell proliferation
          milk ejection
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          negative regulation of wound healing
          cellular response to mechanical stimulus
          atrial cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          negative regulation of DNA biosynthetic process
          regulation of bicellular tight junction assembly
          positive regulation of behavioral fear response
          signal transducer activity
          receptor binding
          gap junction channel activity
          protein binding
          ion transmembrane transporter activity
          SH3 domain binding
          PDZ domain binding
          beta-tubulin binding
          connexin binding
          gap junction channel activity involved in cardiac conduction electrical coupling
          scaffold protein binding
          gap junction channel activity involved in cell communication by electrical coupling

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0