Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPC3 SIRNA Assays ›
          Gene Symbol:
          GPC3
          Gene Name:
          glypican 3
          Gene Aliases:
          DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
          Chromosome Location:
          Chr.X: 133535745 - 133985646 on Build GRCh38
          UniGene ID:
          Hs.644108
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5795

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GPC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164617.1 NP_001158089.1 3 810
          NM_001164618.1 NP_001158090.1 3 762
          NM_001164619.1 NP_001158091.1 2 648
          NM_004484.3 NP_004475.1 3 810
          XM_017029413.1 XP_016884902.1 3 781
          GenBank mRNA AK222761.1 - 3 758
          AK222766.1 - 3 758
          AK300168.1 - 3 733
          AK310196.1 - 2 477
          AK310689.1 - 4 829
          BC035972.1 - 3 758 4603748
          DQ349136.1 - 2 451
          DQ349137.1 - 3 613
          DQ349138.1 - 3 613
          JQ943686.1 - 3 613
          L47125.1 - 3 764
          L47176.1 - 3 759
          U50410.1 - 3 757
          Z37987.1 - 3 721

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_016697, AF185614, AK168402, BC036126, AK159394, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          branching involved in ureteric bud morphogenesis
          glycosaminoglycan biosynthetic process
          glycosaminoglycan catabolic process
          anatomical structure morphogenesis
          anterior/posterior axis specification
          body morphogenesis
          negative regulation of peptidase activity
          glycosaminoglycan metabolic process
          bone mineralization
          osteoclast differentiation
          lung development
          positive regulation of BMP signaling pathway
          embryonic hindlimb morphogenesis
          positive regulation of protein catabolic process
          positive regulation of endocytosis
          negative regulation of smoothened signaling pathway
          positive regulation of smoothened signaling pathway
          negative regulation of growth
          positive regulation of glucose import
          negative regulation of epithelial cell proliferation
          coronary vasculature development
          mesenchymal cell proliferation involved in ureteric bud development
          mesonephric duct morphogenesis
          cell proliferation involved in metanephros development
          negative regulation of canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway, planar cell polarity pathway
          protein binding
          heparan sulfate proteoglycan binding
          peptidyl-dipeptidase inhibitor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bjzzw:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0