Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 15 | 2820 | ||
NM_001292009.1 | NP_001278938.1 | 15 | 2820 | |||
GenBank mRNA | FJ416331.1 | - | 15 | 2828 | ||
FM209056.1 | - | 15 | 2772 |