Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003303.1 | XP_016858792.1 | 2 | 224 | ||
XM_017003304.1 | XP_016858793.1 | 2 | 224 |