siRNA Details
Gene Symbol: UBTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143975.1 | NP_001137447.1 | 1 | 686 |

Gene Symbol: UBTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143975.1 | NP_001137447.1 | 1 | 686 |