Search Thermo Fisher Scientific
Gene Symbol: TRIM49D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206627.1 | NP_001193556.1 | 3 | 691 | ||
XM_011542816.2 | XP_011541118.1 | 4 | 807 | |||
GenBank mRNA | JF968447.1 | - | 1 | 28 | ||
JF968448.1 | - | 1 | 28 |
Gene Symbol: TRIM49D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105522.1 | NP_001098992.1 | 4 | 805 |