Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP53 SIRNA Assays ›
          Gene Symbol:
          TP53
          Gene Name:
          tumor protein p53
          Gene Aliases:
          BCC7, LFS1, P53, TRP53
          Chromosome Location:
          Chr.17: 7668402 - 7687550 on Build GRCh38
          UniGene ID:
          Hs.437460
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s605
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP53
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000546.5 NP_000537.3 7, 8 995
          NM_001126112.2 NP_001119584.1 7, 8 992
          NM_001126113.2 NP_001119585.1 7, 8 995
          NM_001126114.2 NP_001119586.1 7, 8 995
          NM_001126115.1 NP_001119587.1 3, 4 675
          NM_001126116.1 NP_001119588.1 3, 4 675
          NM_001126117.1 NP_001119589.1 3, 4 675
          NM_001126118.1 NP_001119590.1 6, 7 1112
          NM_001276695.1 NP_001263624.1 7, 8 995
          NM_001276696.1 NP_001263625.1 7, 8 995
          NM_001276697.1 NP_001263626.1 3, 4 675
          NM_001276698.1 NP_001263627.1 3, 4 675
          NM_001276699.1 NP_001263628.1 3, 4 675
          NM_001276760.1 NP_001263689.1 7, 8 995
          NM_001276761.1 NP_001263690.1 7, 8 992
          GenBank mRNA AB082923.1 - 7, 8 856
          AF307851.1 - 7, 8 928
          AK223026.1 - 1, 2 88
          AK225838.1 - 7, 8 933
          AK297462.1 - 8, 9 860
          AK297927.1 - 8, 9 905
          AK312568.1 - 7, 8 937
          AM076970.1 - 6, 7 760
          AM076972.1 - 6, 7 793
          AY429684.1 - 6, 7 793
          BC003596.1 - 7, 8 913 3544714
          BT019622.1 - 6, 7 793
          DQ186648.1 - 7, 8 925
          DQ186649.1 - 7, 8 925
          DQ186650.1 - 3, 4 671
          DQ186651.1 - 3, 4 671
          DQ186652.1 - 3, 4 671
          DQ191317.1 - 7, 8 982
          DQ286964.1 - 7, 8 927
          EF101867.1 - 6, 7 760
          EF101868.1 - 6, 7 793
          FJ207420.1 - 7, 8 928
          JN214348.1 - 6, 7 838
          JN601513.1 - 6, 7 793
          JN601514.1 - 6, 7 793
          JN601515.1 - 6, 7 793
          JN601516.1 - 7, 8 767
          JN900492.1 - 6, 7 1101
          JQ026207.1 - 6, 7 783
          JQ026208.1 - 6, 7 788
          JQ026209.1 - 6, 7 798
          JQ694049.1 - 6, 7 816
          JQ694050.1 - 6, 7 816
          JQ694051.1 - 6, 7 816
          KC820708.1 - 3, 4 288
          KC820709.1 - 3, 4 288
          KC820710.1 - 3, 4 288
          KC820711.1 - 3, 4 288
          KC820712.1 - 3, 4 288
          KC820713.1 - 3, 4 288
          KC820714.1 - 3, 4 288
          KC820715.1 - 3, 4 288
          KC820716.1 - 3, 4 288
          KC820717.1 - 3, 4 288
          KC820718.1 - 3, 4 288
          KC820719.1 - 3, 4 288
          KC820720.1 - 3, 4 288
          KC820721.1 - 3, 4 288
          KC820722.1 - 3, 4 288
          KC820723.1 - 3, 4 288
          KC820724.1 - 3, 4 288
          KC820725.1 - 3, 4 288
          KC820726.1 - 3, 4 288
          KC820727.1 - 3, 4 288
          KC820728.1 - 3, 4 288
          KC820729.1 - 3, 4 288
          KC820730.1 - 3, 4 288
          KC820731.1 - 3, 4 288
          KC820732.1 - 3, 4 288
          KC820733.1 - 3, 4 288
          KC820734.1 - 3, 4 288
          KC820735.1 - 3, 4 288
          KC820736.1 - 3, 4 288
          KC820737.1 - 3, 4 288
          KC820738.1 - 3, 4 288
          KC820740.1 - 3, 4 288
          KC820741.1 - 3, 4 288
          KC820742.1 - 3, 4 288
          KC820743.1 - 3, 4 288
          KC820744.1 - 3, 4 288
          KC820745.1 - 3, 4 288
          KC820746.1 - 3, 4 288
          KC820747.1 - 3, 4 288
          KC820748.1 - 3, 4 288
          KC820749.1 - 3, 4 288
          KC820750.1 - 3, 4 288
          KC820751.1 - 3, 4 288
          KC820752.1 - 3, 4 288
          KC820753.1 - 3, 4 288
          KC820754.1 - 3, 4 288
          KC820755.1 - 3, 4 288
          KC820756.1 - 3, 4 288
          KC820757.1 - 3, 4 288
          KC820758.1 - 3, 4 288
          KC820759.1 - 2, 3 177
          KC820760.1 - 3, 4 288
          KC820761.1 - 3, 4 288
          KC820762.1 - 3, 4 288
          KC820763.1 - 3, 4 288
          KC820764.1 - 3, 4 288
          KC820765.1 - 3, 4 288
          KC820766.1 - 3, 4 288
          KC820767.1 - 3, 4 288
          KC820768.1 - 3, 4 288
          KC820769.1 - 3, 4 288
          KC820770.1 - 3, 4 288
          KC820771.1 - 3, 4 288
          KC820772.1 - 3, 4 288
          KC820773.1 - 3, 4 288
          KC820774.1 - 3, 4 288
          KC820775.1 - 3, 4 288
          KC820776.1 - 3, 4 288
          KC820777.1 - 3, 4 288
          KC820778.1 - 3, 4 288
          KC820779.1 - 3, 4 288
          KC820780.1 - 3, 4 288
          KC820781.1 - 3, 4 288
          KC820782.1 - 3, 4 288
          KC820783.1 - 3, 4 288
          KC820784.1 - 3, 4 288
          KC820785.1 - 3, 4 288
          KC820786.1 - 3, 4 288
          M14694.1 - 7, 8 918
          M14695.1 - 7, 8 914
          X01405.1 - 4, 5 494
          X02469.1 - 7, 8 928
          X60011.1 - 6, 7 793
          X60012.1 - 6, 7 793
          X60013.1 - 6, 7 793
          X60014.1 - 6, 7 793
          X60015.1 - 6, 7 793
          X60016.1 - 6, 7 793
          X60017.1 - 6, 7 793
          X60018.1 - 6, 7 793
          X60019.1 - 6, 7 793
          X60020.1 - 6, 7 793

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_003749209.

          Matching Taqman Gene Expression Assay:


          Hs00153340_m1

          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA strand renaturation
          base-excision repair
          nucleotide-excision repair
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein complex assembly
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          ER overload response
          cell cycle arrest
          Ras protein signal transduction
          multicellular organism development
          cell aging
          protein localization
          cell proliferation
          negative regulation of cell proliferation
          determination of adult lifespan
          response to X-ray
          response to gamma radiation
          positive regulation of gene expression
          viral process
          protein sumoylation
          cell differentiation
          negative regulation of cell growth
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of histone deacetylation
          chromatin assembly
          mitotic G1 DNA damage checkpoint
          positive regulation of protein oligomerization
          cellular response to UV
          cellular response to drug
          cellular response to glucose starvation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          entrainment of circadian clock by photoperiod
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of neuron apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          response to antibiotic
          positive regulation of protein export from nucleus
          regulation of mitochondrial membrane permeability
          negative regulation of fibroblast proliferation
          circadian behavior
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of helicase activity
          protein tetramerization
          negative regulation of telomerase activity
          positive regulation of thymocyte apoptotic process
          positive regulation of cell cycle arrest
          cellular response to hypoxia
          cellular response to ionizing radiation
          intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of release of cytochrome c from mitochondria
          replicative senescence
          oxidative stress-induced premature senescence
          intrinsic apoptotic signaling pathway
          oligodendrocyte apoptotic process
          positive regulation of execution phase of apoptosis
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          regulation of cell cycle G2/M phase transition
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          positive regulation of reactive oxygen species metabolic process
          positive regulation of intrinsic apoptotic signaling pathway
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protease binding
          p53 binding
          DNA binding
          chromatin binding
          damaged DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          copper ion binding
          protein binding
          ATP binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          receptor tyrosine kinase binding
          ubiquitin protein ligase binding
          histone acetyltransferase binding
          identical protein binding
          sequence-specific DNA binding
          protein self-association
          transcription regulatory region DNA binding
          protein heterodimerization activity
          protein N-terminus binding
          chaperone binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0