Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105371095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023975.1 | XP_016879464.1 | 1 | 1255 | ||
XM_017023976.1 | XP_016879465.1 | 1 | 1255 |
Gene Symbol: LOC105376747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023973.1 | XP_016879462.1 | 1 | 1254 | ||
XM_017023974.1 | XP_016879463.1 | 1 | 1254 |