Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRM3 SIRNA Assays ›
          Gene Symbol:
          CHRM3
          Gene Name:
          cholinergic receptor muscarinic 3
          Gene Aliases:
          EGBRS, HM3, PBS
          Chromosome Location:
          Chr.1: 239386565 - 239915450 on Build GRCh38
          UniGene ID:
          Hs.155736
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s60844

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000740.2 NP_000731.1 5 2199
          XM_005273032.3 XP_005273089.1 6 2504
          XM_011544041.2 XP_011542343.1 7 2603
          XM_011544043.2 XP_011542345.1 9 2936
          XM_011544044.2 XP_011542346.1 8 2707
          XM_011544045.2 XP_011542347.1 8 2832
          XM_011544046.2 XP_011542348.1 6 2540
          XM_011544047.2 XP_011542349.1 6 2476
          XM_017000152.1 XP_016855641.1 8 5529
          XM_017000153.1 XP_016855642.1 7 2607
          XM_017000154.1 XP_016855643.1 6 2301
          XM_017000155.1 XP_016855644.1 7 2644
          XM_017000156.1 XP_016855645.1 7 2608
          XM_017000157.1 XP_016855646.1 5 2209
          XM_017000158.1 XP_016855647.1 5 2441
          XM_017000159.1 XP_016855648.1 5 2413
          XM_017000160.1 XP_016855649.1 4 3263
          XM_017000161.1 XP_016855650.1 6 2545
          XM_017000162.1 XP_016855651.1 5 2377
          XM_017000163.1 XP_016855652.1 4 6239
          GenBank mRNA AF385589.1 - 1 1420
          AF498917.1 - 1 1420
          AK074575.1 - 1 1577
          BC096844.1 - 1 1421
          BC121026.2 - 2 1563 40117429

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of vascular smooth muscle contraction
          cellular protein modification process
          smooth muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled acetylcholine receptor signaling pathway
          phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway
          G-protein coupled acetylcholine receptor signaling pathway
          synaptic transmission, cholinergic
          nervous system development
          cell proliferation
          positive regulation of smooth muscle contraction
          saliva secretion
          phosphatidylinositol phospholipase C activity
          receptor activity
          protein binding
          drug binding
          G-protein coupled acetylcholine receptor activity
          acetylcholine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0