Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD36B SIRNA Assays ›
Gene Symbol:
ANKRD36B
ANKRD36
Gene Name:
ankyrin repeat domain 36B
ankyrin repeat domain 36
Gene Aliases:
KIAA1641
UNQ2430
Chromosome Location:
Chr.2: 97492663 - 97590245 on Build GRCh38
Chromosome Location:
Chr.2: 97113186 - 97264521 on Build GRCh38
UniGene ID:
Hs.532921
Hs.541894
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s60964
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD36B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025190.3
NP_079466.3
40
4037
XM_005263995.4
XP_005264052.1
40
4317
XM_006712661.1
XP_006712724.1
40
4317
XM_017004578.1
XP_016860067.1
40
4314
XM_017004579.1
XP_016860068.1
38
4215
XM_017004580.1
XP_016860069.1
38
4215
XM_017004581.1
XP_016860070.1
38
4215
XM_017004582.1
XP_016860071.1
38
4215
XM_017004583.1
XP_016860072.1
39
4179
XM_017004584.1
XP_016860073.1
34
4011
XM_017004585.1
XP_016860074.1
38
4072
XM_017004586.1
XP_016860075.1
35
3058
XM_017004587.1
XP_016860076.1
39
4246
XM_017004588.1
XP_016860077.1
38
4111
XM_017004589.1
XP_016860078.1
37
4004
XM_017004590.1
XP_016860079.1
36
3931
XM_017004591.1
XP_016860080.1
22
4035
XM_017004592.1
XP_016860081.1
41
4361
XM_017004593.1
XP_016860082.1
39
4167
XM_017004594.1
XP_016860083.1
38
4060
XM_017004595.1
XP_016860084.1
39
4116
GenBank mRNA
AF432208.1
-
2
407
AF432209.1
-
2
504
AK024934.1
-
2
149
AK056042.1
-
2
266
BC038460.1
-
2
223
BC125132.1
-
2
149
BC125133.1
-
2
149
DB105321.1
-
2
196
Gene Symbol: ANKRD36
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164315.1
NP_001157787.1
72
5765
XM_006712514.3
XP_006712577.1
72
5997
XM_011511130.1
XP_011509432.1
72
5997
XM_017004009.1
XP_016859498.1
72
5997
XM_017004010.1
XP_016859499.1
72
5994
XM_017004011.1
XP_016859500.1
70
5895
XM_017004012.1
XP_016859501.1
70
5895
XM_017004013.1
XP_016859502.1
70
5895
XM_017004014.1
XP_016859503.1
70
5895
XM_017004015.1
XP_016859504.1
70
5895
XM_017004016.1
XP_016859505.1
70
5895
XM_017004017.1
XP_016859506.1
70
5895
XM_017004018.1
XP_016859507.1
70
5895
XM_017004019.1
XP_016859508.1
70
5895
XM_017004020.1
XP_016859509.1
70
5895
XM_017004021.1
XP_016859510.1
70
5895
XM_017004022.1
XP_016859511.1
70
5895
XM_017004023.1
XP_016859512.1
70
5895
XM_017004024.1
XP_016859513.1
71
5859
XM_017004025.1
XP_016859514.1
68
5793
XM_017004026.1
XP_016859515.1
68
5793
XM_017004027.1
XP_016859516.1
68
5793
XM_017004028.1
XP_016859517.1
68
5793
XM_017004029.1
XP_016859518.1
68
5793
XM_017004030.1
XP_016859519.1
68
5793
XM_017004031.1
XP_016859520.1
68
5793
XM_017004032.1
XP_016859521.1
68
5793
XM_017004033.1
XP_016859522.1
68
5793
XM_017004034.1
XP_016859523.1
68
5793
XM_017004035.1
XP_016859524.1
68
5793
XM_017004036.1
XP_016859525.1
68
5793
XM_017004037.1
XP_016859526.1
68
5793
XM_017004038.1
XP_016859527.1
68
5793
XM_017004039.1
XP_016859528.1
66
5691
XM_017004040.1
XP_016859529.1
66
5691
XM_017004041.1
XP_016859530.1
66
5691
XM_017004042.1
XP_016859531.1
66
5691
XM_017004043.1
XP_016859532.1
66
5691
XM_017004044.1
XP_016859533.1
64
5589
XM_017004045.1
XP_016859534.1
64
5589
XM_017004046.1
XP_016859535.1
70
5752
XM_017004047.1
XP_016859536.1
69
5684
XM_017004048.1
XP_016859537.1
68
5611
XM_017004049.1
XP_016859538.1
70
5786
XM_017004050.1
XP_016859539.1
55
4190
XM_017004051.1
XP_016859540.1
53
4404
GenBank mRNA
AB046861.1
-
14
2663
AF543495.1
-
4
1206
AK090926.1
-
2
857
BC146835.1
-
14
2674
8860491
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0